siRNA Details
| Gene Symbol: Has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045020.1 | - | 1 | 2442 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB293662.2 | - | 1 | 364 | Chr. 15 - 56691948 - 56692461 [-] on Build GRCm38 | |

Has2
Genomic Map