siRNA Details
Gene Symbol: Arl4aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016131.1 | - | 2 | 324 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715735.1 | - | 2 | 324 | Chr. 12 - 40038330 - 40040027 [+] on Build GRCm38 |
