siRNA Details
| Gene Symbol: 4930555G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016130.1 | - | 2, 3 | 237 | ||
| BY715733.1 | - | 2, 3 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930555G21Rik| Gene Symbol: 4930555G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016130.1 | - | 2, 3 | 237 | ||
| BY715733.1 | - | 2, 3 | 237 | |||