siRNA Details
| Gene Symbol: E130102H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040708.1 | 2 | 674 | |||
| GenBank mRNA | AK021368.1 | - | 2 | 636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130102H24Rik| Gene Symbol: E130102H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040708.1 | 2 | 674 | |||
| GenBank mRNA | AK021368.1 | - | 2 | 636 | ||