siRNA Details
| Gene Symbol: Nctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002452.2 | 1, 2 | 192 | |||
| GenBank mRNA | AI594936.1 | - | 1, 2 | 192 | ||
| AK035596.1 | - | 1, 2 | 191 | |||
| AK036720.1 | - | 1, 2 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nctc1| Gene Symbol: Nctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002452.2 | 1, 2 | 192 | |||
| GenBank mRNA | AI594936.1 | - | 1, 2 | 192 | ||
| AK035596.1 | - | 1, 2 | 191 | |||
| AK036720.1 | - | 1, 2 | 192 | |||