siRNA Details
| Gene Symbol: Gm20063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045049.1 | 1 | 271 | |||
| GenBank mRNA | AK043502.1 | - | 1 | 272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20063| Gene Symbol: Gm20063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045049.1 | 1 | 271 | |||
| GenBank mRNA | AK043502.1 | - | 1 | 272 | ||