siRNA Details
| Gene Symbol: Gm20063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045049.1 | 1 | 718 | |||
| GenBank mRNA | AK043502.1 | - | 1 | 719 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB261931.2 | - | 1 | 146 | Chr. 12 - 61499675 - 61521248 [-] on Build GRCm38 | |

Gm20063
Genomic Map