siRNA Details
| Gene Symbol: E130101A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021363.1 | - | 1 | 48 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ555165.1 | - | 1 | 272 | Chr. 13 - 81726675 - 81727267 [+] on Build GRCm38 | |

E130101A01Rik
Genomic Map