siRNA Details
| Gene Symbol: E130101A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021363.1 | - | 1 | 502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130101A01Rik| Gene Symbol: E130101A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021363.1 | - | 1 | 502 | ||