| GenBankmRNA | 
						BY091215.1 | 
						- | 
						
							
								3
							 | 
							
								230
							 | 
						
						 | 
						Chr. 7 - 45571157 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI116742.1 | 
						- | 
						
							
								3
							 | 
							
								231
							 | 
						
						 | 
						Chr. 7 - 45571043 - 45575159 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB949865.1 | 
						- | 
						
							
								3
							 | 
							
								262
							 | 
						
						 | 
						Chr. 7 - 45568249 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI182559.1 | 
						- | 
						
							
								3
							 | 
							
								228
							 | 
						
						 | 
						Chr. 7 - 45568347 - 45575156 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BF234373.1 | 
						- | 
						
							
								
							 | 
							
								221
							 | 
						
						 | 
						Chr.  -  -  [-] on  Build  | 
						
					
				
					
						| GenBankmRNA | 
						BI329313.1 | 
						- | 
						
							
								2
							 | 
							
								218
							 | 
						
						 | 
						Chr. 7 - 45567999 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BF659895.1 | 
						- | 
						
							
								2
							 | 
							
								197
							 | 
						
						 | 
						Chr. 7 - 45571173 - 45574776 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						W07892.1 | 
						- | 
						
							
								
							 | 
							
								246
							 | 
						
						 | 
						Chr.  -  -  [-] on  Build  | 
						
					
				
					
						| GenBankmRNA | 
						BF233971.1 | 
						- | 
						
							
								
							 | 
							
								428
							 | 
						
						 | 
						Chr.  -  -  [-] on  Build  | 
						
					
				
					
						| GenBankmRNA | 
						AA241021.1 | 
						- | 
						
							
								2
							 | 
							
								200
							 | 
						
						 | 
						Chr. 7 - 45574455 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA895719.1 | 
						- | 
						
							
								2
							 | 
							
								115
							 | 
						
						 | 
						Chr. 7 - 45570988 - 45574685 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB952915.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45568271 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AW475520.1 | 
						- | 
						
							
								3
							 | 
							
								218
							 | 
						
						 | 
						Chr. 7 - 45570517 - 45575146 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BF385555.1 | 
						- | 
						
							
								
							 | 
							
								220
							 | 
						
						 | 
						Chr.  -  -  [-] on  Build  | 
						
					
				
					
						| GenBankmRNA | 
						CA468575.1 | 
						- | 
						
							
								3
							 | 
							
								296
							 | 
						
						 | 
						Chr. 7 - 45568135 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BB660942.1 | 
						- | 
						
							
								3
							 | 
							
								246
							 | 
						
						 | 
						Chr. 7 - 45570881 - 45575173 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BI247179.1 | 
						- | 
						
							
								2
							 | 
							
								206
							 | 
						
						 | 
						Chr. 7 - 45567987 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB601067.1 | 
						- | 
						
							
								3
							 | 
							
								231
							 | 
						
						 | 
						Chr. 7 - 45568312 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA469521.1 | 
						- | 
						
							
								2
							 | 
							
								202
							 | 
						
						 | 
						Chr. 7 - 45571116 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA388456.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45571063 - 45575146 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB948202.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45568167 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA511853.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45571055 - 45575148 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BI331312.1 | 
						- | 
						
							
								2
							 | 
							
								215
							 | 
						
						 | 
						Chr. 7 - 45568078 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA895654.1 | 
						- | 
						
							
								2
							 | 
							
								194
							 | 
						
						 | 
						Chr. 7 - 45571139 - 45574773 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BF784518.1 | 
						- | 
						
							
								2
							 | 
							
								217
							 | 
						
						 | 
						Chr. 7 - 45568103 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AW258975.1 | 
						- | 
						
							
								3
							 | 
							
								228
							 | 
						
						 | 
						Chr. 7 - 45571122 - 45575156 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BI146741.1 | 
						- | 
						
							
								2
							 | 
							
								284
							 | 
						
						 | 
						Chr. 7 - 45568104 - 45574846 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BF237302.1 | 
						- | 
						
							
								3
							 | 
							
								218
							 | 
						
						 | 
						Chr. 7 - 45568330 - 45575146 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA105155.1 | 
						- | 
						
							
								2
							 | 
							
								220
							 | 
						
						 | 
						Chr. 7 - 45571065 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB949658.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45568155 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI286451.1 | 
						- | 
						
							
								
							 | 
							
								226
							 | 
						
						 | 
						Chr.  -  -  [-] on  Build  | 
						
					
				
					
						| GenBankmRNA | 
						CB950990.1 | 
						- | 
						
							
								3
							 | 
							
								225
							 | 
						
						 | 
						Chr. 7 - 45568233 - 45575153 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB948045.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45568205 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI930329.1 | 
						- | 
						
							
								3
							 | 
							
								228
							 | 
						
						 | 
						Chr. 7 - 45571017 - 45575156 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI746710.1 | 
						- | 
						
							
								3
							 | 
							
								228
							 | 
						
						 | 
						Chr. 7 - 45574073 - 45575156 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BY091214.1 | 
						- | 
						
							
								3
							 | 
							
								230
							 | 
						
						 | 
						Chr. 7 - 45571157 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA250638.1 | 
						- | 
						
							
								3
							 | 
							
								248
							 | 
						
						 | 
						Chr. 7 - 45574412 - 45575176 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA473150.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45570985 - 45575146 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AA239239.1 | 
						- | 
						
							
								2
							 | 
							
								201
							 | 
						
						 | 
						Chr. 7 - 45570985 - 45574779 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						BB845898.1 | 
						- | 
						
							
								3
							 | 
							
								230
							 | 
						
						 | 
						Chr. 7 - 45571076 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB599533.1 | 
						- | 
						
							
								3
							 | 
							
								269
							 | 
						
						 | 
						Chr. 7 - 45570187 - 45575194 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CA481621.1 | 
						- | 
						
							
								3
							 | 
							
								266
							 | 
						
						 | 
						Chr. 7 - 45568145 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AW259999.1 | 
						- | 
						
							
								3
							 | 
							
								228
							 | 
						
						 | 
						Chr. 7 - 45568333 - 45575155 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						CB593529.1 | 
						- | 
						
							
								3
							 | 
							
								232
							 | 
						
						 | 
						Chr. 7 - 45568277 - 45575157 [-] on  Build GRCm38 | 
						
					
				
					
						| GenBankmRNA | 
						AI049249.1 | 
						- | 
						
							
								3
							 | 
							
								217
							 | 
						
						 | 
						Chr. 7 - 45570999 - 45575145 [-] on  Build GRCm38 |