siRNA Details
| Gene Symbol: 1700014B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005972.1 | - | 1 | 694 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV040851.2 | - | 1 | 87 | Chr. 2 - 6322265 - 6322463 [-] on Build GRCm38 | |

1700014B07Rik
Genomic Map