siRNA Details
| Gene Symbol: 3010001F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045451.1 | 4 | 712 | |||
| NR_045452.1 | 5 | 693 | ||||
| GenBank mRNA | AK013876.1 | - | 4 | 715 | ||
| EL960655.1 | - | 2 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3010001F23Rik| Gene Symbol: 3010001F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045451.1 | 4 | 712 | |||
| NR_045452.1 | 5 | 693 | ||||
| GenBank mRNA | AK013876.1 | - | 4 | 715 | ||
| EL960655.1 | - | 2 | 284 | |||