siRNA Details
| Gene Symbol: 2310069B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040520.1 | 4 | 1808 | |||
| GenBank mRNA | AK078808.1 | - | 4 | 1809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310069B03Rik| Gene Symbol: 2310069B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040520.1 | 4 | 1808 | |||
| GenBank mRNA | AK078808.1 | - | 4 | 1809 | ||