siRNA Details
| Gene Symbol: 9330109K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020364.1 | - | 1 | 352 | ||
| BB180676.1 | - | 1 | 206 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721268.1 | - | 1 | 352 | Chr. 8 - 56129641 - 56130070 [+] on Build GRCm38 | |

9330109K16Rik
Genomic Map