siRNA Details
| Gene Symbol: 1700080N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040500.1 | 2, 3 | 298 | |||
| GenBank mRNA | AK006958.1 | - | 2, 3 | 299 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707011.1 | - | 2, 3 | 299 | Chr. 2 - 4132065 - 4141141 [-] on Build GRCm38 | |

1700080N15Rik
Genomic Map