siRNA Details
| Gene Symbol: 1700080N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040500.1 | 2 | 263 | |||
| GenBank mRNA | AK006958.1 | - | 2 | 264 | ||
| AK016735.1 | - | 3 | 1063 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700080N15Rik| Gene Symbol: 1700080N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040500.1 | 2 | 263 | |||
| GenBank mRNA | AK006958.1 | - | 2 | 264 | ||
| AK016735.1 | - | 3 | 1063 | |||