siRNA Details
Gene Symbol: 1700080N15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040500.1 | 1 | 73 | |||
GenBank mRNA | AK006958.1 | - | 1 | 74 | ||
AK016735.1 | - | 1 | 71 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700080N15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040500.1 | 1 | 73 | |||
GenBank mRNA | AK006958.1 | - | 1 | 74 | ||
AK016735.1 | - | 1 | 71 |