siRNA Details
Gene Symbol: 2410004I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037963.1 | 2 | 244 | |||
GenBank mRNA | AK010390.1 | - | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2410004I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037963.1 | 2 | 244 | |||
GenBank mRNA | AK010390.1 | - | 2 | 246 |