siRNA Details
| Gene Symbol: 2410004I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037963.1 | 3 | 908 | |||
| GenBank mRNA | AK010390.1 | - | 3 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410004I01Rik| Gene Symbol: 2410004I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037963.1 | 3 | 908 | |||
| GenBank mRNA | AK010390.1 | - | 3 | 910 | ||