siRNA Details
| Gene Symbol: 2410004I01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037963.1 | 3 | 592 | |||
| GenBank mRNA | AK010390.1 | - | 3 | 594 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY709833.1 | - | 3 | 594 | Chr. 11 - 102958354 - 102964154 [-] on Build GRCm38 | |

2410004I01Rik
Genomic Map