siRNA Details
| Gene Symbol: 1700052M09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006774.1 | - | 1 | 290 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706757.1 | - | 1 | 293 | Chr. 14 - 29840855 - 29841725 [+] on Build GRCm38 | |

1700052M09Rik
Genomic Map