siRNA Details
| Gene Symbol: Gm36584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076849.1 | - | 3 | 669 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV265541.2 | - | 3 | 585 | Chr. 7 - 82852498 - 82866992 [-] on Build GRCm38 | |

Gm36584
Genomic Map