siRNA Details
| Gene Symbol: Zfp626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020189.1 | - | 1 | 191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE651548.1 | - | 1 | 73 | Chr. 7 - 27819859 - 27820416 [+] on Build GRCm38 | |

Zfp626
Genomic Map