siRNA Details
| Gene Symbol: Gm17116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041104.1 | - | 2 | 138 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637544.1 | - | 2 | 135 | Chr. 14 - 64111636 - 64114639 [-] on Build GRCm38 | |

Gm17116
Genomic Map