siRNA Details
| Gene Symbol: B230119M05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045454.1 | 2 | 53 | |||
| GenBank mRNA | AK020983.1 | - | 2 | 54 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727074.1 | - | 2 | 54 | Chr. X - 134684869 - 134686462 [-] on Build GRCm38 | |

B230119M05Rik
Genomic Map