siRNA Details
| Gene Symbol: A930041H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020950.1 | - | 1 | 899 | ||
| Gene Symbol: Parp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046177.1 | - | 1 | 119 | ||

A930041H05Rik
Genomic Map