siRNA Details
| Gene Symbol: 1110007E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003532.1 | - | 1 | 772 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV007945.1 | - | 1 | 161 | Chr. 4 - 124640533 - 124640837 [+] on Build GRCm38 | |

1110007E10Rik
Genomic Map