siRNA Details
Gene Symbol: U90926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033483.1 | 4 | 250 | |||
GenBank mRNA | U90926.1 | - | 4 | 261 | ||
W13227.1 | - | 2 | 347 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: U90926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033483.1 | 4 | 250 | |||
GenBank mRNA | U90926.1 | - | 4 | 261 | ||
W13227.1 | - | 2 | 347 |