siRNA Details
| Gene Symbol: U90926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033483.1 | 5 | 287 | |||
| GenBank mRNA | U90926.1 | - | 5 | 298 | ||
| W13227.1 | - | 3 | 384 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

U90926| Gene Symbol: U90926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033483.1 | 5 | 287 | |||
| GenBank mRNA | U90926.1 | - | 5 | 298 | ||
| W13227.1 | - | 3 | 384 | |||