siRNA Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036940.1 | - | 1 | 603 | ||
| AK043267.1 | - | 1 | 452 | |||
| AK172512.1 | - | 1 | 545 | |||
| AK172537.1 | - | 1 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mef2c| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036940.1 | - | 1 | 603 | ||
| AK043267.1 | - | 1 | 452 | |||
| AK172512.1 | - | 1 | 545 | |||
| AK172537.1 | - | 1 | 540 | |||