siRNA Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036940.1 | - | 1 | 1389 | ||
AK043267.1 | - | 1 | 1236 | |||
AK172512.1 | - | 1 | 1331 | |||
AK172537.1 | - | 1 | 1326 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036940.1 | - | 1 | 1389 | ||
AK043267.1 | - | 1 | 1236 | |||
AK172512.1 | - | 1 | 1331 | |||
AK172537.1 | - | 1 | 1326 |