siRNA Details
| Gene Symbol: 1700121L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045453.1 | 3 | 532 | |||
| GenBank mRNA | AK007232.1 | - | 3 | 533 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707323.1 | - | 3 | 533 | Chr. X - 104816776 - 104842455 [+] on Build GRCm38 | |

1700121L16Rik
Genomic Map