siRNA Details
| Gene Symbol: 1700122D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007236.1 | - | 2 | 376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700122D07Rik| Gene Symbol: 1700122D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007236.1 | - | 2 | 376 | ||