siRNA Details
| Gene Symbol: 1700121N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036593.1 | 4 | 674 | |||
| GenBank mRNA | AK007234.1 | - | 4 | 709 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700121N20Rik| Gene Symbol: 1700121N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036593.1 | 4 | 674 | |||
| GenBank mRNA | AK007234.1 | - | 4 | 709 | ||