siRNA Details
| Gene Symbol: Gm13807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040529.1 | 5 | 1051 | |||
| GenBank mRNA | AK029608.1 | - | 5 | 1053 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV260387.2 | - | 2 | 407 | Chr. 2 - 93416095 - 93417830 [+] on Build GRCm38 | |

Gm13807
Genomic Map