siRNA Details
Gene Symbol: Gm13807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040529.1 | 6 | 1234 | |||
GenBank mRNA | AK029608.1 | - | 6 | 1236 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260387.2 | - | 3 | 590 | Chr. 2 - 93416095 - 93417830 [+] on Build GRCm38 |
