siRNA Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085332.1 | - | 1 | 1020 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB498209.2 | - | 1 | 319 | Chr. 9 - 52072544 - 52073185 [+] on Build GRCm38 | |

Rdx
Genomic Map