siRNA Details
| Gene Symbol: Gm40811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK154198.1 | - | 1 | 835 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY763754.1 | - | 1 | 225 | Chr. 10 - 117149290 - 117149979 [+] on Build GRCm38 | |

Gm40811
Genomic Map