siRNA Details
| Gene Symbol: Gm8541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136239.1 | - | 5 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720319.1 | - | 5 | 512 | Chr. 2 - 63528005 - 63575288 [-] on Build GRCm38 | |

Gm8541
Genomic Map