siRNA Details
| Gene Symbol: Gm11476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077405.1 | - | 4 | 1662 | ||
| AV293622.2 | - | 1 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11476| Gene Symbol: Gm11476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077405.1 | - | 4 | 1662 | ||
| AV293622.2 | - | 1 | 267 | |||