siRNA Details
| Gene Symbol: Gm34016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038089.1 | - | 1 | 83 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB631454.1 | - | 1 | 83 | Chr. 12 - 73490762 - 73508670 [-] on Build GRCm38 | |

Gm34016
Genomic Map