siRNA Details
| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037997.1 | 2 | 192 | |||
| GenBank mRNA | AK136726.1 | - | 2 | 195 | ||
| BU743775.1 | - | 216 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310015D24Rik| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037997.1 | 2 | 192 | |||
| GenBank mRNA | AK136726.1 | - | 2 | 195 | ||
| BU743775.1 | - | 216 | ||||