siRNA Details
| Gene Symbol: Kcnq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080597.1 | - | 1 | 1587 | ||
| BC059899.1 | - | 1 | 1796 | |||
| BC096016.1 | - | 1 | 1796 | |||
| CA320890.1 | - | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq2| Gene Symbol: Kcnq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080597.1 | - | 1 | 1587 | ||
| BC059899.1 | - | 1 | 1796 | |||
| BC096016.1 | - | 1 | 1796 | |||
| CA320890.1 | - | 1 | 95 | |||