siRNA Details
| Gene Symbol: Gm4719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045952.1 | 4 | 701 | |||
| GenBank mRNA | AK131717.1 | - | 4 | 702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4719| Gene Symbol: Gm4719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045952.1 | 4 | 701 | |||
| GenBank mRNA | AK131717.1 | - | 4 | 702 | ||