siRNA Details
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013401.1 | - | 1 | 611 | ||
AK170452.1 | - | 2 | 2786 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712394.1 | - | 1 | 607 | Chr. 5 - 105730678 - 105731477 [+] on Build GRCm38 |
