siRNA Details
| Gene Symbol: Lrrc8d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013401.1 | - | 1 | 178 | ||
| AK170452.1 | - | 2 | 2353 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712394.1 | - | 1 | 174 | Chr. 5 - 105730678 - 105731477 [+] on Build GRCm38 | |

Lrrc8d
Genomic Map