siRNA Details
| Gene Symbol: 2610307P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045053.1 | 9 | 1781 | |||
| NR_045054.1 | 8 | 1694 | ||||
| GenBank mRNA | AK038887.1 | - | 3 | 554 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632643.1 | - | 3 | 554 | Chr. 13 - 28460681 - 28521108 [-] on Build GRCm38 | |

2610307P16Rik
Genomic Map