siRNA Details
| Gene Symbol: 2610307P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045053.1 | 9 | 2338 | |||
| NR_045054.1 | 8 | 2251 | ||||
| GenBank mRNA | AK038887.1 | - | 3 | 1111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610307P16Rik| Gene Symbol: 2610307P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045053.1 | 9 | 2338 | |||
| NR_045054.1 | 8 | 2251 | ||||
| GenBank mRNA | AK038887.1 | - | 3 | 1111 | ||