siRNA Details
| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038885.1 | - | 1845 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230072E10Rik| Gene Symbol: A230072E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038885.1 | - | 1845 | |||