siRNA Details
| Gene Symbol: 1700030F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045731.1 | 2, 3 | 182 | |||
| GenBank mRNA | AK006540.1 | - | 2, 3 | 183 | ||
| BY706558.1 | - | 2, 3 | 183 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV043825.2 | - | 1, 2 | 52 | Chr. 6 - 117679211 - 117751057 [-] on Build GRCm38 | |

1700030F04Rik
Genomic Map